Methyl benzenesulphonate 98%

Supplier: Ambeed
Danger

A221783-10G A221783-25G A221783-5G
77165-240EA 7 USD
77165-240 77165-242 77165-244
Methyl benzenesulphonate 98%
Methyl benzenesulphonate

Precaución: for Research Use Only

Formula: C₇H₈O₃S
MDL Number: MFCD00014737
CAS Number: 80-18-2
UN: 3265
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR