4-Methoxy-1-naphthalenemethanol 98%

Supplier: Ambeed
Danger

A221314-1G A221314-250MG A221314-5G
77192-890EA 39 USD
77192-890 77192-892 77192-894
4-Methoxy-1-naphthalenemethanol 98%
4-Methoxy-1-naphthalenemethanol

Precaución: for Research Use Only

Formula: C₁₂H₁₂O₂
MDL Number: MFCD04089264
CAS Number: 16820-54-5
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR