2,6-Diiodopyridine 95%

Supplier: Ambeed
Warning

A220922-250MG A220922-1G A220922-5G A220922-100MG
77165-212EA 36 USD
77165-212 77568-130 77568-132 77892-349
2,6-Diiodopyridine 95%
2,6-Diiodopyridine

Precaución: For research use only.

Formula: C₅H₃I₂N
MDL Number: MFCD07366728
CAS Number: 53710-17-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR