16-Mercaptopalmitic acid 98%

Supplier: Ambeed
Danger

A220863-100MG A220863-250MG A220863-1G A220863-5G
77192-884EA 44 USD
77192-884 77192-886 77593-432 77746-311
16-Mercaptopalmitic acid 98%
16-Mercaptopalmitic acid

Precaución: for Research Use Only

Formula: C₁₆H₃₂O₂S
MDL Number: MFCD00674018
CAS Number: 69839-68-5
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR