Methyl 2-methyl-1H-indole-6-carboxylate 95%

Supplier: Ambeed
Warning

A220810-100MG A220810-1G A220810-250MG
77165-206EA 60.77 USD
77165-206 77165-208 77165-210
Methyl 2-methyl-1H-indole-6-carboxylate 95%
Methyl 2-methyl-1H-indole-6-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₁NO₂
MW: 189.21 g/mol
MDL Number: MFCD18381247
CAS Number: 184150-96-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR