17-AAG 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
17-(Allylamino)-17-demethoxygeldanamycin, Tanespimycin, (4E,6Z,8S,9S,10E,12S,13R,14S,16R)-19-(Allylamino)-13-hydroxy-8,14-dimethoxy-4,10,12,16-tetramethyl-3,20,22-trioxo-2-azabicyclo[16.3.1]docosa-1(21),4,6,10,18-pentaen-9-yl carbamate
A220462-10MG
A220462-1MG
77192-880EA
71.07
USD
77192-880
77192-882
17-AAG 98%
17-AAG
Precaución: for Research Use Only
Formula:
C₃₁H₄₃N₃O₈ MW: 585.7 g/mol |
MDL Number:
MFCD04973892 CAS Number: 75747-14-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...