1,2-Epoxydodecane 95%

Supplier: Ambeed
Danger

A220457-100G A220457-1G A220457-25G A220457-5G
77192-874EA 176.13 USD
77192-874 77192-876 77192-878 77746-310
1,2-Epoxydodecane 95%
1,2-Epoxydodecane

Precaución: for Research Use Only

Formula: C₁₂H₂₄O
MW: 184.32 g/mol
MDL Number: MFCD00005150
CAS Number: 2855-19-8
UN: 3082
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR