3-Amino-6-bromo-1H-indazole 98%

Supplier: Ambeed
Warning

Synonyms: 6-Bromo-1H-indazol-3-amine

A219894-100G A219894-10G A219894-1G A219894-250MG A219894-5G A219894-25G
77165-168EA 584.01 USD
77165-168 77165-170 77165-172 77165-174 77165-176 77683-635
3-Amino-6-bromo-1H-indazole 98%
3-Amino-6-bromo-1H-indazole

Precaución: For research use only.

Formula: C₇H₆BrN₃
MW: 212.05 g/mol
MDL Number: MFCD05665872
CAS Number: 404827-77-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR