4-Bromo-2-ethylthiazole 95%

Supplier: Ambeed
Warning

A219232-100MG A219232-1G
77165-158EA 85.49 USD
77165-158 77165-160
4-Bromo-2-ethylthiazole 95%
4-Bromo-2-ethylthiazole

Precaución: for Research Use Only

Formula: C₅H₆BrNS
MW: 192.08 g/mol
MDL Number: MFCD11223239
CAS Number: 120258-27-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR