2-Bromo-1-(phenylsulfonyl)-1H-pyrrolo[2,3-b]pyridine 97%

Supplier: Ambeed
Warning

A218959-100MG A218959-1G A218959-250MG
77192-850EA 75.99 USD
77192-850 77192-852 77192-854
2-Bromo-1-(phenylsulfonyl)-1H-pyrrolo[2,3-b]pyridine 97%
2-Bromo-1-(phenylsulfonyl)-1H-pyrrolo[2,3-b]pyridine

Precaución: for Research Use Only

Formula: C₁₃H₉BrN₂O₂S
MDL Number: MFCD16875828
CAS Number: 1227271-03-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR