5-(Methoxymethyl)-1H-pyrazol-3-amine 98%

Supplier: Ambeed
Danger

A218820-100MG A218820-1G A218820-250MG
77165-148EA 88.58 USD
77165-148 77165-150 77165-152
5-(Methoxymethyl)-1H-pyrazol-3-amine 98%
5-(Methoxymethyl)-1H-pyrazol-3-amine

Precaución: for Research Use Only

Formula: C₅H₉N₃O
MW: 127.14 g/mol
MDL Number: MFCD22576523
CAS Number: 739366-03-1
UN: 3259
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR