(Z)-8-dodecen-1-ylacetate 90%

Supplier: Ambeed
Danger

A218268-1G A218268-250MG
77192-830EA 178 USD
77192-830 77746-302
(Z)-8-dodecen-1-ylacetate 90%
(Z)-8-dodecen-1-ylacetate

Precaución: for Research Use Only

Formula: C₁₄H₂₆O₂
MW: 226.35 g/mol
CAS Number: 28079-04-1
UN: 3082
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR