5-Chloro-2-methoxy-4-pyridineboronic acid 95%

Supplier: Ambeed
Warning

A217914-100MG A217914-1G A217914-250MG A217914-25G A217914-5G
77165-146EA 5 USD
77165-146 77892-341 77892-342 77892-343 77892-344
5-Chloro-2-methoxy-4-pyridineboronic acid 95%
5-Chloro-2-methoxy-4-pyridineboronic acid

Precaución: for Research Use Only

Formula: C₆H₇BClNO₃
MDL Number: MFCD06658398
CAS Number: 475275-69-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR