1-Phenylcyclopropanamine hydrochloride 97%

Supplier: Ambeed
Warning

A217881-1G A217881-250MG A217881-5G
77165-140EA 53.56 USD
77165-140 77165-142 77165-144
1-Phenylcyclopropanamine hydrochloride 97%
1-Phenylcyclopropanamine hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₂ClN
MW: 169.6500 g/mol
MDL Number: MFCD07995722
CAS Number: 73930-39-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR