tert-Butyl (3-hydroxypyridin-2-yl)carbamate 97%

Supplier: Ambeed
Warning

A217578-1G A217578-5G
77192-814EA 9 USD
77192-814 77192-816 77192-818
tert-Butyl (3-hydroxypyridin-2-yl)carbamate 97%
tert-Butyl (3-hydroxypyridin-2-yl)carbamate

Precaución: for Research Use Only

Formula: C₁₀H₁₄N₂O₃
MDL Number: MFCD07781139
CAS Number: 902835-93-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR