3-Chloro-2-fluoroisonicotinaldehyde 95%

Supplier: Ambeed
Warning

A217475-100MG A217475-1G
77165-130EA 59.74 USD
77165-130 77165-132
3-Chloro-2-fluoroisonicotinaldehyde 95%
3-Chloro-2-fluoroisonicotinaldehyde

Precaución: for Research Use Only

Formula: C₆H₃ClFNO
MW: 159.546 g/mol
MDL Number: MFCD12756634
CAS Number: 1149587-02-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR