1,4-Dibenzyloxy-2-nitrobenzene 97%

Supplier: Ambeed
Warning

Synonyms: (((2-Nitro-1,4-phenylene)bis(oxy))bis(methylene))dibenzene

A217300-10G A217300-1G A217300-5G
77192-808EA 120 USD
77192-808 77192-810 77192-812
1,4-Dibenzyloxy-2-nitrobenzene 97%
1,4-Dibenzyloxy-2-nitrobenzene

Precaución: for Research Use Only

Formula: C₂₀H₁₇NO₄
MDL Number: MFCD00082778
CAS Number: 51792-85-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR