6-Chloro-1H-pyrazolo[4,3-c]pyridine-3-carboxylic acid 98%
Supplier: Ambeed
|
Warning
|
A216702-1G
A216702-5G
A216702-250MG
77165-118EA
209.09
USD
77165-118
77165-120
77683-624
6-Chloro-1H-pyrazolo[4,3-c]pyridine-3-carboxylic acid 98%
6-Chloro-1H-pyrazolo[4,3-c]pyridine-3-carboxylic acid
Precaución: For research use only.
Formula:
C₇H₄ClN₃O₂ MW: 197.57 g/mol |
MDL Number:
MFCD11042721 CAS Number: 1314903-71-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...