5-Fluoro-o-toluidine 98%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-4-fluorotoluene, 5-Fluoro-2-methylaniline

A216394-100G A216394-10G A216394-25G A216394-500G
77192-796EA 42.23 USD
77192-796 77192-798 77192-800 77192-802
5-Fluoro-o-toluidine 98%
5-Fluoro-o-toluidine

Precaución: for Research Use Only

Formula: C₇H₈FN
MW: 125.15 g/mol
MDL Number: MFCD00007764
CAS Number: 367-29-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR