2'-Chloro-4'-nitroacetophenone 98%

Supplier: Ambeed
Warning

Synonyms: 1-(2-Chloro-4-nitrophenyl)ethanone

A216060-1G A216060-250MG A216060-25G A216060-5G
77192-788EA 21 USD
77192-788 77192-790 77192-792 77192-794
2'-Chloro-4'-nitroacetophenone 98%
2'-Chloro-4'-nitroacetophenone

Precaución: for Research Use Only

Formula: C₈H₆ClNO₃
MW: 199.59 g/mol
MDL Number: MFCD00017340
CAS Number: 67818-41-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR