1,1,1-Trifluoro-6-phenylhex-5-ene-2,4-dione 97%

Supplier: Ambeed
Warning

A215984-1G A215984-250MG A215984-5G
77192-782EA 62 USD
77192-782 77192-784 77192-786
1,1,1-Trifluoro-6-phenylhex-5-ene-2,4-dione 97%
1,1,1-Trifluoro-6-phenylhex-5-ene-2,4-dione

Precaución: for Research Use Only

Formula: C₁₂H₉F₃O₂
MDL Number: MFCD00760722
CAS Number: 18931-64-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR