[2'-(Amino-κN)[1,1'-biphenyl]-2-yl-κC][[5-(diphenylphosphino)-9,9-dimethyl-9H-xanthen-4-yl]diphenylphosphine-κP](methanesulfonato-κO)palladium 98%

Supplier: Ambeed
Warning

A214875-100MG A214875-1G A214875-5G A214875-250MG A214875-25G
77165-082EA 7 USD
77165-082 77165-084 77550-388 77593-394 77892-336
[2'-(Amino-κN)[1,1'-biphenyl]-2-yl-κC][[5-(diphenylphosphino)-9,9-dimethyl-9H-xanthen-4-yl]diphenylphosphine-κP](methanesulfonato-κO)palladium 98%
[2'-(Amino-κN)[1,1'-biphenyl]-2-yl-κC][[5-(diphenylphosphino)-9,9-dimethyl-9H-xanthen-4-yl]diphenylphosphine-κP](methanesulfonato-κO)palladium

Precaución: for Research Use Only

Formula: C₅₂H₄₅NO₄P₂PdS
MDL Number: MFCD22572675
CAS Number: 1445085-97-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR