N-(3-(Dimethylamino)propyl)-4-(8-hydroxyquinolin-6-yl)benzamide 98%

Supplier: Ambeed
Warning

A214803-10MG A214803-5MG A214803-100MG A214803-250MG A214803-50MG
77165-076EA 68.99 USD
77165-076 77165-078 77165-080 77892-333 77892-334 77892-335
N-(3-(Dimethylamino)propyl)-4-(8-hydroxyquinolin-6-yl)benzamide 98%
N-(3-(Dimethylamino)propyl)-4-(8-hydroxyquinolin-6-yl)benzamide

Precaución: for Research Use Only

Formula: C₂₁H₂₃N₃O₂
MDL Number: MFCD28053518
CAS Number: 1222800-79-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR