6-Bromo-2-iodopyridin-3-ol 95%

Supplier: Ambeed
Warning

A214667-250MG A214667-1G
77165-074EA 32.96 USD
77165-074 77683-610
6-Bromo-2-iodopyridin-3-ol 95%
6-Bromo-2-iodopyridin-3-ol

Precaución: For research use only.

Formula: C₅H₃BrINO
MW: 299.89 g/mol
MDL Number: MFCD11977407
CAS Number: 188057-35-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR