5-Bromo-1-methyl-4-nitro-1H-pyrazole 95%

Supplier: Ambeed
Warning

A214659-100MG A214659-1G A214659-250MG A214659-5G
77165-066EA 36.05 USD
77165-066 77165-068 77165-070 77165-072
5-Bromo-1-methyl-4-nitro-1H-pyrazole 95%
5-Bromo-1-methyl-4-nitro-1H-pyrazole

Precaución: for Research Use Only

Formula: C₄H₄BrN₃O₂
MW: 205.997 g/mol
MDL Number: MFCD00195044
CAS Number: 89607-13-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR