(S)-4-(5-Chloro-2-(isopropylamino)pyridin-4-yl)-N-(1-(3-chlorophenyl)-2-hydroxyethyl)-1H-pyrrole-2-carboxamide hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: Ulixertinib hydrochloride

A214234-10MG A214234-1MG A214234-25MG A214234-50MG A214234-1G
77192-748EA 28.84 USD
77192-748 77192-750 77192-752 77192-754 77746-295
(S)-4-(5-Chloro-2-(isopropylamino)pyridin-4-yl)-N-(1-(3-chlorophenyl)-2-hydroxyethyl)-1H-pyrrole-2-carboxamide hydrochloride 98%
(S)-4-(5-Chloro-2-(isopropylamino)pyridin-4-yl)-N-(1-(3-chlorophenyl)-2-hydroxyethyl)-1H-pyrrole-2-carboxamide hydrochloride

Precaución: for Research Use Only

Formula: C₂₁H₂₃Cl₃N₄O₂
MW: 469.79 g/mol
MDL Number: MFCD22543853
CAS Number: 1956366-10-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR