8-Bromo-3-fluoroquinoline 95%

Supplier: Ambeed

A214224-100MG A214224-250MG A214224-50MG
77165-050EA 149 USD
77165-050 77165-052 77165-054
8-Bromo-3-fluoroquinoline 95%
8-Bromo-3-fluoroquinoline

Precaución: for Research Use Only

Formula: C₉H₅BrFN
MDL Number: MFCD12546390
CAS Number: 834884-06-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR