4-Propenylveratrole 98%

Supplier: Ambeed
Warning

Synonyms: 4-(1-Propenyl)pyrocatechol dimethyl ether, 4-(1-Propenyl)-1,2-dimethoxybenzene, 1,2-Dimethoxy-4-Propenylbenzene, 1,2-Dimethoxy-4-(1-propenyl)benzene, Methyl isoeugenol, O-Methyl Isoeugenol

A214096-100G A214096-10G A214096-25G A214096-500G A214096-5G A214096-1G
77192-738EA 30.9 USD
77192-738 77192-740 77192-742 77192-744 77192-746 77746-293
4-Propenylveratrole 98%
4-Propenylveratrole

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₂
MW: 178.23 g/mol
MDL Number: MFCD00009282
CAS Number: 93-16-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR