1-(2-Hydroxyethyl)-2-pyrrolidone ≥98%

Supplier: Ambeed
Warning

A213977-100G A213977-25G
77192-726EA 36 USD
77192-726 77192-728
1-(2-Hydroxyethyl)-2-pyrrolidone ≥98%
1-(2-Hydroxyethyl)-2-pyrrolidone

Precaución: for Research Use Only

Formula: C₆H₁₁NO₂
MDL Number: MFCD00014103
CAS Number: 3445-11-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR