2-(3,5-Difluorophenyl)propan-2-amine hydrochloride 98%

Supplier: Ambeed
Warning

A213483-100MG A213483-10G A213483-250MG A213483-5G A213483-1G
77165-038EA 16.48 USD
77165-038 77165-040 77165-042 77165-044 77683-607
2-(3,5-Difluorophenyl)propan-2-amine hydrochloride 98%
2-(3,5-Difluorophenyl)propan-2-amine hydrochloride

Precaución: For research use only.

Formula: C₉H₁₁F₂N·ClH
MW: 207.648 g/mol
MDL Number: MFCD19687116
CAS Number: 1063733-81-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR