3-Bromo-2-fluorobenzamide 98%

Supplier: Ambeed
Danger

A213459-5G A213459-250MG
77165-030EA 120 USD
77165-030 77892-327
3-Bromo-2-fluorobenzamide 98%
3-Bromo-2-fluorobenzamide

Precaución: for Research Use Only

Formula: C₇H₅BrFNO
MDL Number: MFCD12913463
CAS Number: 871353-25-2
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR