tert-Butyl (6-amino-6-oxohexyl)carbamate 95%

Supplier: Ambeed
Warning

A213212-1G A213212-250MG A213212-5G
77165-024EA 72.1 USD
77165-024 77165-026 77165-028
tert-Butyl (6-amino-6-oxohexyl)carbamate 95%
tert-Butyl (6-amino-6-oxohexyl)carbamate

Precaución: for Research Use Only

Formula: C₁₁H₂₂N₂O₃
MW: 230.30 g/mol
MDL Number: MFCD24466641
CAS Number: 85535-56-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR