3-((Benzyloxy)methyl)oxetane 95%

Supplier: Ambeed
Warning

A213111-100MG A213111-250MG
77165-020EA 29.87 USD
77165-020 77165-022
3-((Benzyloxy)methyl)oxetane 95%
3-((Benzyloxy)methyl)oxetane

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₂
MW: 178.23 g/mol
MDL Number: MFCD20482348
CAS Number: 1003013-76-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR