1-Chloro-2-iodo-3-nitrobenzene 97%

Supplier: Ambeed
Warning

A213033-1G A213033-250MG A213033-25G A213033-5G
77165-012EA 32.96 USD
77165-012 77165-014 77165-016 77165-018
1-Chloro-2-iodo-3-nitrobenzene 97%
1-Chloro-2-iodo-3-nitrobenzene

Precaución: for Research Use Only

Formula: C₆H₃ClINO₂
MW: 283.45 g/mol
MDL Number: MFCD08275977
CAS Number: 32337-97-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR