2-Bromo-4,5-dichloroanisole 97%

Supplier: Ambeed
Warning

Synonyms: 1-Bromo-4,5-dichloro-2-methoxybenzene

A212846-1G A212846-250MG A212846-5G
77192-686EA 185.4 USD
77192-686 77192-688 77192-690
2-Bromo-4,5-dichloroanisole 97%
2-Bromo-4,5-dichloroanisole

Precaución: for Research Use Only

Formula: C₇H₅BrCl₂O
MW: 255.9240 g/mol
MDL Number: MFCD11845923
CAS Number: 174913-15-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR