4-Chloro-1-ethynyl-2-fluorobenzene 95%

Supplier: Ambeed
Danger

A212771-1G A212771-250MG A212771-5G
77165-002EA 235.87 USD
77165-002 77165-004 77683-605
4-Chloro-1-ethynyl-2-fluorobenzene 95%
4-Chloro-1-ethynyl-2-fluorobenzene

Precaución: For research use only.

Formula: C₈H₄ClF
MW: 154.56 g/mol
MDL Number: MFCD08703530
CAS Number: 188472-71-1
UN: 1325
ADR: 4.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR