Zaltoprofen 97%

Supplier: Ambeed
Warning

Synonyms: 2-(10,11-Dihydro-10-oxodibenzo[b,f]thiepin-2-yl)propionic Acid, 2-(10-Oxo-10,11-dihydrodibenzo[b,f]thiepin-2-yl)propanoic acid

A212638-10G A212638-1G A212638-25G
77192-680EA 51.5 USD
77192-680 77192-682 77192-684
Zaltoprofen 97%
Zaltoprofen

Precaución: for Research Use Only

Formula: C₁₇H₁₄O₃S
MW: 298.36 g/mol
MDL Number: MFCD00864323
CAS Number: 74711-43-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR