1-Methylcyclopropanecarboxylic acid 95%

Supplier: Ambeed
Danger

A212533-1G A212533-250MG A212533-25G A212533-5G A212533-10G A212533-100G
77164-988EA 11 USD
77164-988 77164-990 77164-992 77164-994 77550-368 77892-325
1-Methylcyclopropanecarboxylic acid 95%
1-Methylcyclopropanecarboxylic acid

Precaución: for Research Use Only

Formula: C₅H₈O₂
MDL Number: MFCD00001290
CAS Number: 6914-76-7
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR