Methyl 3-(4-hydroxyphenyl)hex-4-ynoate 95%

Supplier: Ambeed
Warning

A212378-100MG A212378-1G A212378-250MG
77164-980EA 33.99 USD
77164-980 77164-982 77164-984
Methyl 3-(4-hydroxyphenyl)hex-4-ynoate 95%
Methyl 3-(4-hydroxyphenyl)hex-4-ynoate

Precaución: for Research Use Only

Formula: C₁₃H₁₄O₃
MW: 218.25 g/mol
MDL Number: MFCD24713927
CAS Number: 865234-02-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR