Histamine dihydrochloride 98%

Supplier: Ambeed
Warning

A211645-100G A211645-10G A211645-25G A211645-500G A211645-50MG
77164-954EA 70 USD
77164-954 77164-956 77164-958 77164-960 77164-962
Histamine dihydrochloride 98%
Histamine dihydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₁Cl₂N₃
MDL Number: MFCD00012703
CAS Number: 56-92-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR