2-[3-(Trifluoromethyl)phenyl]propan-2-amine hydrochloride 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-(3-(Trifluoromethyl)phenyl)propan-2-amine hydrochloride
A211618-100MG
A211618-1G
A211618-250MG
77164-948EA
22.66
USD
77164-948
77164-950
77164-952
2-[3-(Trifluoromethyl)phenyl]propan-2-amine hydrochloride 97%
2-[3-(Trifluoromethyl)phenyl]propan-2-amine hydrochloride
Precaución: for Research Use Only
Formula:
C₁₀H₁₂F₃N·ClH MW: 239.665 g/mol |
MDL Number:
MFCD14708148 CAS Number: 1439900-32-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...