1-(3-Aminopropyl)pyrrolidine 97%

Supplier: Ambeed
Danger

Synonyms: N-(3-Aminopropyl)pyrrolidine, 3-(Pyrrolidin-1-yl)propan-1-amine

A211399-1G A211399-25G A211399-5G A211399-10G
77192-652EA 9.27 USD
77192-652 77192-654 77192-656 77568-040
1-(3-Aminopropyl)pyrrolidine 97%
1-(3-Aminopropyl)pyrrolidine

Precaución: For research use only.

Formula: C₇H₁₆N₂
MW: 128.22 g/mol
MDL Number: MFCD00014100
CAS Number: 23159-07-1
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR