3-Chloro-5-nitrophenol 97%

Supplier: Ambeed
Danger

A211344-1G A211344-250MG A211344-10G A211344-25G A211344-5G
77164-944EA 17 USD
77164-944 77164-946 77892-321 77892-322 77892-323
3-Chloro-5-nitrophenol 97%
3-Chloro-5-nitrophenol

Precaución: for Research Use Only

Formula: C₆H₄ClNO₃
MDL Number: MFCD15976952
CAS Number: 618-63-3
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR