L-Allylglycine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(S)-(-)-alpha-Allylglycine, H-Ally-Gly-OH, H-Gly(Ally)-OH, 2-Allyl-L-glycine, (S)-(-)-2-Amino-4-pentenoic acid, (2S)-2-Aminopent-4-enoic acid, L-2-Amino-4-pentenoic acid L-Allylglycine L-alpha-Allylglycine, (S)-2-Allylglycine
A211329-10G
A211329-1G
A211329-250MG
A211329-25G
A211329-5G
A211329-100G
77164-934EA
51
USD
77164-934
77164-936
77164-938
77164-940
77164-942
77612-082
L-Allylglycine 98%
L-Allylglycine
Precaución: for Research Use Only
Formula:
C₅H₉NO₂ MW: 115.132 g/mol |
MDL Number:
MFCD00002627 CAS Number: 16338-48-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...