3-Methylbenzamidine hydrochloride 97%
Supplier: Ambeed
|
Warning
|
A211128-1G
A211128-250MG
77192-648EA
17
USD
77192-648
77192-650
3-Methylbenzamidine hydrochloride 97%
3-Methylbenzamidine hydrochloride
Precaución: for Research Use Only
Formula:
C₈H₁₁ClN₂ |
MDL Number:
MFCD02180878 CAS Number: 20680-59-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...