XL388 98%

Supplier: Ambeed
Warning

Synonyms: (7-(6-Aminopyridin-3-yl)-2,3-dihydrobenzo[f][1,4]oxazepin-4(5H)-yl)(3-fluoro-2-methyl-4-(methylsulphonyl)phenyl)methanone

A210267-10MG A210267-1MG A210267-25MG A210267-50MG A210267-5MG
77192-608EA 70.04 USD
77192-608 77192-610 77192-612 77192-614 77192-616
XL388 98%
XL388

Precaución: for Research Use Only

Formula: C₂₃H₂₂FN₃O₄S
MW: 455.5 g/mol
MDL Number: MFCD24386875
CAS Number: 1251156-08-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR