2-(Pyridin-4-yl)acetic acid hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: 4-Pyridylacetic acid hydrochloride

A210184-100G A210184-10G A210184-1G A210184-25G A210184-5G
77192-598EA 237.93 USD
77192-598 77192-600 77192-602 77192-604 77192-606
2-(Pyridin-4-yl)acetic acid hydrochloride 98%
2-(Pyridin-4-yl)acetic acid hydrochloride

Precaución: for Research Use Only

Formula: C₇H₈ClNO₂
MW: 173.60 g/mol
MDL Number: MFCD00012827
CAS Number: 6622-91-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR