GNE-7915 ≥98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(4-((4-(Ethylamino)-5-(trifluoromethyl)pyrimidin-2-yl)amino)-2-fluoro-5-methoxyphenyl)(morpholino)methanone, [4-[[4-(ethylamino)-5-(trifluoromethyl)pyrimidin-2-yl]amino]-2-fluoro-5-methoxyphenyl]-morpholin-4-ylmethanone
A210153-10MG
A210153-1MG
A210153-50MG
A210153-5MG
77192-590EA
36.05
USD
77192-590
77192-592
77192-594
77192-596
GNE-7915 ≥98%
GNE-7915
Precaución: for Research Use Only
Formula:
C₁₉H₂₁F₄N₅O₃ MW: 443.4 g/mol |
MDL Number:
MFCD25977005 CAS Number: 1351761-44-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...