2,7-Dibromo-9-hexylcarbazole 97%

Supplier: Ambeed
Warning

Synonyms: 2,7-Dibromo-9-hexyl-9H-carbazole

A209595-100G A209595-1G A209595-25G A209595-5G
77192-576EA 553.11 USD
77192-576 77192-578 77192-580 77192-582
2,7-Dibromo-9-hexylcarbazole 97%
2,7-Dibromo-9-hexylcarbazole

Precaución: for Research Use Only

Formula: C₁₈H₁₉Br₂N
MW: 409.17 g/mol
MDL Number: MFCD29089359
CAS Number: 654676-12-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR