(S)-1-Methylpiperidine-2-carboxylic acid 95%
Supplier: Ambeed
|
Warning
|
A209142-100MG
A209142-250MG
A209142-1G
A209142-5G
77164-894EA
16
USD
77164-894
77164-896
77593-358
77892-310
(S)-1-Methylpiperidine-2-carboxylic acid 95%
(S)-1-Methylpiperidine-2-carboxylic acid
Precaución: for Research Use Only
Formula:
C₇H₁₃NO₂ |
MDL Number:
MFCD19228347 CAS Number: 41447-18-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...